missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-FLT3 (Tyr842) Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5105168
Description
Antibody detects endogenous levels of Flt3 / CD135 only when phosphorylated at Tyr842.
FLT3 is a tyrosine protein kinase that is a member of the growth factor receptor tyrosine kinase family. The receptor is activated by binding of the fms-related tyrosine kinase 3 ligand to the extracellular domain, which induces homodimer formation in the plasma membrane leading to autophosphorylation of the receptor. The activated receptor kinase subsequently phosphorylates and activates multiple cytoplasmic effector molecules in pathways involved in apoptosis, proliferation, and differentiation of hematopoietic cells in bone marrow. Mutations that result in the constitutive activation of this receptor result in acute myeloid leukemia and acute lymphoblastic leukemia.Chemical Identifiers
| Phospho-FLT3 (Tyr842) | |
| -20°C | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat, Monkey | |
| FLT3 | |
| B230315G04; CD135; CD135 antigen; fetal liver kinase 2; Fetal liver kinase-2; FL cytokine receptor; FLK2; Flk-2; FLT3; Flt-3; fms related tyrosine kinase 3; FMS-like tyrosine kinase 3; fms-related tyrosine kinase 3; growth factor receptor tyrosine kinase type III; Ly72; OTTHUMP00000214829; Receptor-type tyrosine-protein kinase FLT3; RP11-153M24.3; stem cell tyrosine kinase 1; STK1; STK-1; tyrosine-protein kinase FLT3; tyrosine-protein kinase receptor flk-2; wmfl | |
| FLT3 | |
| Primary | |
| Sequential chromatography |
| 1 mg/mL | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P36888, Q00342 | |
| 140635, 14255, 2322 | |
| A synthesized peptide derived from human FLT3(Accession P36888), corresponding to amino acid residues around phosphorylated Tyr842. | |
| Antibody |
Specifications
| Phospho-FLT3 (Tyr842) | |
| Polyclonal | |
| Unconjugated | |
| FLT3 | |
| B230315G04; CD135; CD135 antigen; fetal liver kinase 2; Fetal liver kinase-2; FL cytokine receptor; FLK2; Flk-2; FLT3; Flt-3; fms related tyrosine kinase 3; FMS-like tyrosine kinase 3; fms-related tyrosine kinase 3; growth factor receptor tyrosine kinase type III; Ly72; OTTHUMP00000214829; Receptor-type tyrosine-protein kinase FLT3; RP11-153M24.3; stem cell tyrosine kinase 1; STK1; STK-1; tyrosine-protein kinase FLT3; tyrosine-protein kinase receptor flk-2; wmfl | |
| Rabbit | |
| Sequential chromatography | |
| RUO | |
| 140635, 14255, 2322 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P36888, Q00342 | |
| FLT3 | |
| A synthesized peptide derived from human FLT3(Accession P36888), corresponding to amino acid residues around phosphorylated Tyr842. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |