missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-ErbB2 (HER-2) (Tyr1248) Recombinant Rabbit Monoclonal Antibody (PSH04-03)

Rabbit Recombinant Monoclonal Antibody

Supplier:  Invitrogen™ MA555601

Catalog No. MA555601


Only null left
Explore available promotions

Description

Description

Positive Control: SK-Br-3 cell lysate, SK-Br-3 starved for 4 hours add 200 ng/mL EGF for 15 minutes cell lysate, HeLa starved for 4 hours add 200 ng/mL EGF for 15 minutes cell lysate, HeLa cells starved for 4 hours add 200 ng/mL EGF for 15 minutes. Tissue Specificity: Low tissue specificity. Subcellular Location: Cell membrane, Cell projection, ruffle membrane; Early endosome, Cytoplasm, perinuclear region, Nucleus. Sequence Similarities: No Cross with Mouse/Rat. Predicted band size: 138 kDa.

ErbB2 (HER2) is a receptor tyrosine kinase that is overexpressed in some breast tumors. Herceptin, used in treatment of metastatic Her2-positive cancer, is monoclonal antibody targeting this kinase.
Specifications

Specifications

Phospho-ErbB2 (HER-2) (Tyr1248)
Recombinant Monoclonal
1 mg/mL
PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
P04626
ERBB2
Synthetic phospho-peptide corresponding to residues surrounding tyr1248 of human her2.
100 μL
Primary
Human
Antibody
IgG
Western Blot, Immunocytochemistry
PSH04-03
Unconjugated
ERBB2
avian erythroblastosis oncogene B 2; Avian erythroblastosis viral (v-erb-B2) oncogene homologue 2 (neuro/glioblastoma derived oncogene homolog); CD antigen CD340; CD340; c-erb B2/neu protein; c-erbB2; C-erbB-2; c-neu; epidermal growth factor receptor-related protein; erbb 2; ERBB2; erb-b2; Erbb-2; ErbB2 (pTyr1139); ErbB2 (pY1139); ErbB2 phospho Y1139; erb-b2 receptor tyrosine kinase 2; her 2; HER2; HER-2; HER-2/neu; herstatin; human epidermal growth factor receptor 2; Kiaa3023; metas; Metastatic lymph node gene 19 protein; mKIAA3023; MLN 19; MLN19; NEU; Neu oncogene; NEU proto-oncogene; neuro/glioblastoma derived oncogene homolog; neuroblastoma/glioblastoma derived oncogene homolog; NGL; p185erbB2; p185neu; phospho ErbB2; proto-oncogene c-ErbB-2; Proto-oncogene Neu; receptor tyrosine-protein kinase erbB-2; TKR1; Tyrosine kinase-type cell surface receptor HER2; v-erb-b2 avian erythroblastic leukemia viral oncogene 2; v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2; v-erb-b2 avian erythroblastic leukemia viral oncoprotein 2; v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastoma derived oncogene homolog; v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastoma derived oncogene homolog (avian)
Rabbit
Protein A
RUO
2064
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions