Learn More
Invitrogen™ Phospho-CREB (Ser133) Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Supplier:  Invitrogen™ PA14619
Catalog No. PA14619
Only null left
Description
Description
This antibody is predicted to react with bovine, canine, chicken, human, mouse, non-human primate, rat, sheep, Xenopus and zebrafish based on 100% sequence homology. This antibody is specific for the ~45 kDa CREB phosphorylated at Ser133 in Western blots of rat brain lysates. Immunolabeling can be blocked by lambda-phosphatase treatment.
CREB (Cyclic AMP response element binding protein) is a 43 kDa basic/leucine zipper transcription factor that binds the cyclic AMP response element (CRE) and activates transcription in response to a variety of extracellular signals including neurotransmitters, hormones, membrane depolarization, and growth and neurotrophic factors. Activation of CREB is dependent upon the phosphorylation of serine 133. Phosphorylation of CREB occurs via p44/42 MAP kinase and p90RSK, and via p38 MAP kinase and MSK1. Although CREB will bind DNA independent of its phosphorylation state, only the phosphorylated form is competent as a transcription factor. CREB binding protein (CBP), a transcriptional coactivator that directly interacts with CREB, binds to CREB in the region of serine 133. CREB is involved in different cellular processes including the synchronization of circadian rhythmicity, differentiation of adipose cells, and learning and memory. In humans, the gene is located on the q arm of chromosome 2. Diseases associated with CREB dysfunction include Alzheimer's Disease, histiocytoma and soft tissue melanoma.
Specifications
Chemical Identifiers
Phospho-CREB (Ser133)
-20°C, Avoid Freeze/Thaw Cycles
Polyclonal
Liquid
IgG
Mouse, Rat
CREB1
2310001E10Rik; 3526402H21Rik; active transcription factor CREB; AV083133; cAMP response element binding protein 1; cAMP responsive element binding protein 1; cAMP-response element-binding protein-1; cAMP-responsive element-binding protein 1; CREB; CREB1; Creb-1; cyclic adenosine 3',5'-monophosphate response element-binding protein CREB; cyclic AMP-responsive element-binding protein 1; htCREB; MGC9284; OTTHUMP00000206661; OTTHUMP00000206662; OTTHUMP00000206667; transactivator protein; Y protein
CREB1, CrebB
Primary
Affinity chromatography
0.25 mg/mL
Immunohistochemistry, Western Blot
Unconjugated
Rabbit
RUO
0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
P15337, Q01147
12912, 81646
Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser133 of rat CREB1 conjugated to KLH.
Antibody
Specifications
Phospho-CREB (Ser133)
Polyclonal
Unconjugated
CREB1
2310001E10Rik; 3526402H21Rik; active transcription factor CREB; AV083133; cAMP response element binding protein 1; cAMP responsive element binding protein 1; cAMP-response element-binding protein-1; cAMP-responsive element-binding protein 1; CREB; CREB1; Creb-1; cyclic adenosine 3',5'-monophosphate response element-binding protein CREB; cyclic AMP-responsive element-binding protein 1; htCREB; MGC9284; OTTHUMP00000206661; OTTHUMP00000206662; OTTHUMP00000206667; transactivator protein; Y protein
Rabbit
Affinity chromatography
RUO
12912, 81646
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry, Western Blot
0.25 mg/mL
0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
P15337, Q01147
CREB1, CrebB
Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser133 of rat CREB1 conjugated to KLH.
100 μL
Primary
Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions