missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-CDK9 (Thr186) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA517197
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517197 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517197 Supplier Invitrogen™ Supplier No. PA517197
Only null left

Rabbit Polyclonal Antibody

It is not recommended to aliquot this antibody.

CDK9 is a serine/threonine kinase which when complexed with CyclinT1 forms an active holoenzyme complex. This complex plays a key role in regulating the eukaryotic cell cycle. CDK9 is coexpressed and copurified with CyclinT1. CDK9 regulates cytokine inducible transcription networks by facilitating promoter recognition of target transcription factors. CDK9 promotes RNA synthesis in genetic programs for cell growth, differentiation, and viral pathogenesis. The CDK9/cyclin-K complex also has kinase activity towards CTD of RNAPII and can substitute for CDK9/cyclin-T P-TEFb in vitro. Chronic activation of CDK9 causes cardiac myocyte enlargement leading to cardiac hypertrophy and confers a predisposition to heart failure.

Specifications

Antigen Phospho-CDK9 (Thr186)
Applications Western Blot
Classification Polyclonal
Concentration 126 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene CDK9
Gene Accession No. P50750, Q641Z4, Q99J95
Gene Alias C-2k; CDC2L4; CDC2-related kinase; Cdk9; Cell division cycle 2-like protein kinase 4; Cell division protein kinase 9; CTK1; cyclin dependent kinase 9; cyclin-dependent kinase 9; cyclin-dependent kinase 9 (CDC2-related kinase); PITALRE; RP11-228B15.5; serine/threonine protein kinase PITALRE; Serine/threonine-protein kinase PITALRE; TAK; Tat-associated kinase complex catalytic subunit
Gene Symbols CDK9
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues surrounding pThr186 of human CDK9.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1025, 107951, 362110
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less