missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-CCR5 (Ser337) Recombinant Mouse Monoclonal Antibody (V14/2)

Mouse Recombinant Monoclonal Antibody

Supplier:  Invitrogen™ MA551824

Catalog No. MA551824


Only null left
Explore available promotions

Description

Description

V14/2 antibody has been demonstrated to exclusively recognise CCR5 (CD195) phosphorylated at Ser 337.

Human immunodeficiency virus (HIV) and related virus require coreceptors, in addition to CD4, to infect target cells. Some G protein-coupled receptors including CCR5, CXCR4, CCR3, CCR2b and CCR8 in the chemokine receptor family, and four new human molecules GPR15, STRL33, GPR1 and V28 were recently identified as HIV coreceptors. Among them, CCR5 (CC-CKR-5) is a principal coreceptor for macrophage- and dual-tropic HIV-1 strains fusion and entry of human white blood cells. CCR5 is required for the infection by HIV-1, HIV-2, and SIV. The beta-chemokines RANTES, MIP-alpha and MIP-beta are the ligands for CCR5 and prevent infection by M-tropic HIV-1. CCR5 associates with the surface CD4-gp120 of HIV complex and leads to membrane fusion and virus entry of target cells. The amino-terminal domain and the extracellular loops of CCR5 serve as HIV biding sites. CCR5 messenger RNA is expressed in lymphoid organs and monocytes.
Specifications

Specifications

Phospho-CCR5 (Ser337)
Recombinant Monoclonal
1 mg/mL
PBS with 0.02% ProClin 300
P51681
CCR5
This antibody was raised by immunizing mice with a synthetic phospho peptide corresponding to the C-Terminal region of human CCR5 conjugated to BSA.
200 μg
Primary
Human
Antibody
IgG2b κ
ELISA, Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry
V14/2
Unconjugated
CCR5
AM4-7; C-C chemokine receptor type 5; C-C CKR-5; C-C motif chemokine receptor 5 (gene/pseudogene); C-C motif chemokine receptor 5 A159A; CCCKR5; CC-CKR-5; Ccr5; CCR-5; CD195; CD195 antigen; chemokine (C-C motif) receptor 5; chemokine (C-C) receptor 5; chemokine receptor CCR5; chemr13; CKR5; CKR-5; Cmkbr5; HIV-1 fusion coreceptor; IDDM22; MIP-1 alpha receptor
Mouse
Protein A
RUO
1234
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions