missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phospho-c-Myc (Ser71) Rabbit anti-Human, Polyclonal, MilliporeSigma™
Description
phospho-c-Myc (Ser71) Polyclonal antibody specifically detects phospho-c-Myc (Ser71) in Human samples. It is validated for Peptide Inhibition Assay, Western Blotting.
Specifications
Specifications
| Antigen | phospho-c-Myc (Ser71) |
| Applications | Inhibition Assays, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.20 mg/mL |
| Conjugate | Unconjugated |
| Formulation | Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. |
| Host Species | Rabbit |
| Immunogen | KLH-conjugated linear peptide corresponding to 11 amino acids surrounding phosphoserine 71 from human c-Myc. |
| Purification Method | Affinity Purified |
| Quantity | 25 μL |
| Show More |