missing translation for 'onlineSavingsMsg'
Learn More

phospho-c-Myc (Ser71) Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABC147225UL
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABC147225UL 25 μL
ABC1472100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABC147225UL Supplier MilliporeSigma Supplier No. ABC147225UL
Only null left

Rabbit Polyclonal Antibody

phospho-c-Myc (Ser71) Polyclonal antibody specifically detects phospho-c-Myc (Ser71) in Human samples. It is validated for Peptide Inhibition Assay, Western Blotting.

Specifications

Antigen phospho-c-Myc (Ser71)
Applications Inhibition Assays, Western Blot
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 11 amino acids surrounding phosphoserine 71 from human c-Myc.
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Stable for 1 year at +2°C to +8°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less