missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-ATRIP (Ser68) Polyclonal Antibody

Description
Phospho-ATRIP (Ser68) Polyclonal Antibody for Western Blot, ICC/IF
Specifications
Specifications
| Antigen | Phospho-ATRIP (Ser68) |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | ATRIP |
| Gene Accession No. | Q8BMG1, Q8WXE1 |
| Gene Alias | 6620401K05Rik; AGS1; ATM and Rad3 related interacting protein; ATM and Rad3-related-interacting protein; ATR interacting protein; ATR IP; ATR-interacting protein; Atrip; DNase III; DRN3; FLJ12343; unnamed protein product |
| Gene Symbols | ATRIP |
| Show More |