missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-4EBP1/2/3 (Thr46, Thr32) Recombinant Rabbit Monoclonal Antibody (23GB3850)
GREENER_CHOICE

Catalog No. MA553650
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553650 100 μL
MA553651 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553650 Supplier Invitrogen™ Supplier No. MA553650
Only null left

Rabbit Recombinant Monoclonal Antibody

Phospho-4EBP1/2/3 (Thr46, Thr32) Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

4EBP1, 4EBP2, and 4EBP3 are members of the eukaryotic translation initiation factor 4E-binding protein (4EBP) family. These proteins play a crucial role in the regulation of protein synthesis, specifically by controlling the availability of the eukaryotic translation initiation factor 4E (eIF4E). 4EBP1, 4EBP2, and 4EBP3 are structurally similar but exhibit some differences in their tissue distribution and expression levels. They are widely expressed in various tissues and cell types and have been implicated in the regulation of different biological processes, including cell growth, proliferation, and survival. Dysregulation of 4EBP proteins has also been associated with various diseases, including cancer.

Specifications

Antigen Phospho-4EBP1/2/3 (Thr46, Thr32)
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB3850
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene EIF4EBP1
Gene Accession No. Q13541, Q60876, Q62622
Gene Alias 4EBP1/2/3; 4E-BP1/2/3; BP-1/2/3; EIF4E-Binding Protein 1/2/3; EIF4EBP1/2/3; Eukaryotic Translation Initiation Factor 4E Binding Protein 1/2/3; Eukaryotic Translation Initiation Factor 4E-Binding Protein 1/2/3; PHAS-I; Phosphorylated Heat- And Acid-Stable Protein Regulated By Insulin 1/2/3
Gene Symbols EIF4EBP1
Host Species Rabbit
Immunogen A synthetic peptide of human Phospho-eIF4EBP1/2/3 (Thr46+Thr46+Thr32) (39-53AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116636, 13685, 1978
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less