missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phosphate Sensor

Catalog No. PV4406
Encompass
Change view
Click to view available options
Quantity:
10 nmol
100 nmol; 10,000 assay(s)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV4406 10 nmol
PV4407 100 nmol; 10,000 assay(s)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PV4406 Supplier Invitrogen™ Supplier No. PV4406
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Phosphate Sensor

The Phosphate Sensor is a highly sensitive reagent for the detection of inorganic phosphate, either in real-time kinetic or end-point mode. This product is a purified form of recombinant E. coli phosphate-binding protein labeled with the fluorophore MDCC, which is sensitive to changes in its environment. Binding of inorganic phosphate by Phosphate Sensor is rapid and tight (Kd ∼ 0.1μM), resulting in a large increase in fluorescence. The Phosphate Sensor detects phosphate in the high-nanomolar to low-micromolar range, which is 100-fold more sensitive than malachite green. This reagent provides you with a sensitive assay for detecting enzymes that produce inorganic phosphate either directly or indirectly through coupled reactions, such as phosphatases, ATPases, GTPases, and phosphodiesterases.

Specifications

Target GTPases, ATPases, Phosphatases, Phosphodiesterases
For Use With (Application) Fluorescence Intensity
Assay Phosphatase Assay
Content And Storage Phosphate Sensor is supplied as a 500 μM solution in 10 mM Tris (pH 7.6) and 50 mM NaCl. Store product at -80°C.
Detection Method Fluorescence
Label or Dye MDCC
Product Type Phosphate Sensor
Quantity 10 nmol
Format 384-well plate
Excitation/Emission 430⁄450
Readout Kinetic
Shipping Condition Dry Ice
For Use With (Equipment) Microplate Reader
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.