missing translation for 'onlineSavingsMsg'
Learn More

Phosphate Buffer Solution, pH 7.5, USP 27 Reagent, Ricca Chemical

Supplier:  Ricca Chemical Company R580825020F

Catalog No. R580825020F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

7778-77-0
USP 27
Odorless
Cubitainer
[K+].OP(O)([O-])=O
136.08
CHEBI:63036
20 L
H2KO4P
MFCD00011401,MFCD00147253
GNSKLFRGEWLPPA-UHFFFAOYSA-M
potassium dihydrogen phosphate
516951

Chemical Identifiers

7778-77-0
136.08
GNSKLFRGEWLPPA-UHFFFAOYSA-M
CHEBI:63036
[K+].OP(O)([O-])=O
H2KO4P
MFCD00011401,MFCD00147253
516951
potassium dihydrogen phosphate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : General

SDS
Documents

Documents

Promotions

Promotions