missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphate buffer solution, pH 7.2, Solstice
Specifications
Specifications
| Chemical Name or Material | Phosphate Buffer Solution Ph 7.2 |
| For Use With (Application) | pH Calibration |
| pH | 7.2 |
| Physical Form | Liquid |
| Quantity | 2 L |
| Solution Type | pH Buffer |
| Packaging | Poly Bottle |
| Grade | Laboratory |