missing translation for 'onlineSavingsMsg'
Learn More

Phosphate Buffer Solution, pH 6.8, for Phenols, Ricca Chemical

Catalog No. 580716
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
580716 500 mL
580732 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 580716 Supplier Ricca Chemical Company Supplier No. 580716
Only null left

For the Determination of Phenols

Specifications

Chemical Name or Material Phosphate Buffer Solution
CAS 10049-21-5
Concentration 1X
For Use With (Application) Water Analysis
pH 7
Physical Form Liquid
Quantity 500 mL
Solution Type Buffer
Packaging Natural Poly Bottle
Grade Laboratory