missing translation for 'onlineSavingsMsg'
Learn More

Phosphate Buffer, pH 7.2, APHA for BOD, Reagents

Catalog No. 01336460
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336460 1 L
01336459 20 L
01336458 4 L
01336457 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01336460 Supplier Labchem Inc Supplier No. C72711501A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Phosphate Buffer, pH 7.2, APHA for BOD, Reagent, use for Biological oxygen demand. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Phosphate Buffer, pH 7.2, APHA for BOD
CAS 7732-18-5
Boiling Point 100°C
Color Colorless
pH 7.2
Physical Form Liquid
Quantity 1 L
Solution Type Buffer
Packaging Poly Bottle
Specific Gravity 1.05
Sterility Non-sterile
Synonym Biological Oxygen Demand Buffer
Grade Reagent
Show More Show Less