missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphate Buffer pH 6.8, for Phenols, LabChem™
Specifications
Specifications
| Chemical Name or Material | Phosphate Buffer |
| Color | Colorless |
| Name Note | For Phenols |
| CAS | 7758-11-4, 7778-77-0 |
| CAS Max % | 82.23, 10.54, 7.23 |
| Molecular Formula | HK2O4P |
| Linear Formula | KH2PO4 |
| Solubility Information | Soluble in water |
| InChI Key | ZPWVASYFFYYZEW-UHFFFAOYSA-L |
| Molecular Weight (g/mol) | 174.17 |
| Show More |