missing translation for 'onlineSavingsMsg'
Learn More

Phosphate Buffer, 50 mM, pH 6.50, Ricca Chemical

Catalog No. R57975004C
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R57975004C 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R57975004C Supplier Ricca Chemical Company Supplier No. R57975004C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ready-to-use phosphate buffer formulated for laboratory applications.

Specifications

Chemical Name or Material Phosphate Buffer
Name Note 50mM - pH 6.50
CAS 7732-18-5, 7778-77-0, 1310-58-3
CAS Min % 97.0, 0.65, 0.15
Color Colorless
CAS Max % 100.0, 0.67, 0.16
Concentration 50 mM
For Use With (Application) Water Analysis
pH 6.5
Physical Form Liquid
Quantity 4 L
Solution Type Buffer
Packaging Amber Glass Bottle
Grade Laboratory
Show More Show Less