missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Phosphate, 0.5M buffer soln., pH 8.0
CAS: 7558-80-7119.976 g/mol
Supplier: Thermo Scientific Chemicals J61722AP
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 7558-80-7 | |
| Liquid | |
| Plastic Bottle |
| 0.5M buffer soln. | |
| 500 mL | |
| 119.976 |
Chemical Identifiers
| 7558-80-7 | |
| 119.976 |
Safety and Handling
Recommended Storage : Ambient Temperatures
RUO – Research Use Only