missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phosphate, 0.5M buffer soln., pH 7.5

CAS: 7558-80-7119.976 g/mol

Supplier:  Thermo Scientific Chemicals J63349AK

 View more versions of this product

Catalog No. AAJ63349AK


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

7558-80-7
Liquid
Plastic Bottle
0.5M buffer soln.
250 mL
119.976

Chemical Identifiers

7558-80-7
119.976
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Ambient Temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only