missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phosphate, 0.2M buffer soln., pH 7.2

CAS: 7558-80-7119.976 g/mol

Supplier:  Thermo Scientific Chemicals J62899K2

 View more versions of this product

Catalog No. AAJ62899K2

Only null left
Explore available promotions

Description

Description

Sodium phosphate monobasic is a reagent with very high buffering capacity widely used in molecular biology, biochemistry and chromatography. Useful in conjunction with Sodium Phosphate, Dibasic in the preparation of biological buffers. Used in many applications including the purification of antibodies. It is also used in HPLC. Phosphates are often used in foods and in water treatment. It is added in animal feed, toothpaste, and evaporated milk. It is used as a thickening agent and emulsifier.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

7558-80-7
Liquid
Plastic Bottle
0.2M buffer soln.
1 L
119.976

Chemical Identifiers

7558-80-7
119.976
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Ambient Temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only