missing translation for 'onlineSavingsMsg'
Learn More

Phosphate, 0.2M buffer soln., pH 6.8, Thermo Scientific Chemicals

Catalog No. aaj60325k2
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60325K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ60325K2 Supplier Thermo Scientific Chemicals Supplier No. J60325K2
Only null left

Widely used in molecular biology, biochemistry and chromatography. Useful in conjunction with Sodium Phosphate, dibasic in the preparation of biological buffers. Used in many applications including the purification of antibodies.

This buffer is widely used in molecular biology, biochemistry and chromatography. Useful in conjunction with Sodium Phosphate, dibasic in the preparation of biological buffers. Used in many applications including the purification of antibodies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7558-80-7
Molecular Weight (g/mol) 119.976

Specifications

Concentration 0.2M buffer soln.
Physical Form Liquid
Quantity 1 L
Packaging Plastic Bottle
Recommended Storage Ambient Temperatures

RUO – Research Use Only