missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phosphate, 0.2M buffer soln., pH 4.4

Catalog No. p-7076544
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60467AP 500 mL
AAJ60467K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60467AP Supplier Thermo Scientific Chemicals Supplier No. J60467AP
Only null left

Widely used in molecular biology, biochemistry and chromatography. Useful in conjunction with Sodium Phosphate, dibasic in the preparation of biological buffers. Used in many applications including the purification of antibodies. | CAS: 7558-80-7119.976 g/mol

This buffer is widely used in molecular biology, biochemistry and chromatography. Useful in conjunction with Sodium Phosphate, dibasic in the preparation of biological buffers. Used in many applications including the purification of antibodies. It is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7558-80-7
Molecular Weight (g/mol) 119.976

Specifications

Concentration 0.2M buffer soln.
Physical Form Liquid
Quantity 500 mL
Packaging Plastic Bottle
Recommended Storage Keep cold

RUO – Research Use Only