missing translation for 'onlineSavingsMsg'
Learn More

Phosphatase, Acid from Wheat Germ, TCI America™

Catalog No. P0251100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0251100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P0251100MG Supplier TCI America Supplier No. P0251100MG
Only null left

Chemical Identifiers

CAS 9001-77-8
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00131847
InChI Key GZCWLCBFPRFLKL-UHFFFAOYSA-N
Synonym Acid Phosphatase
PubChem CID 12951370
IUPAC Name 1-prop-2-ynoxypropan-2-ol
SMILES CC(COCC#C)O

Specifications

Quantity 100 mg
Physical Form Crystalline Powder
Chemical Name or Material Phosphatase, Acid from Wheat Germ
TSCA Yes
Recommended Storage Refrigerator