accessibility menu, dialog, popup

UserName

CAS RN 9001-77-8

H3COCHOH

CAS RN 9001-77-8

IUPAC Name: 1-(prop-2-yn-1-yloxy)propan-2-ol
Synonyms: 1-(prop-2-yn-1-yloxy)propan-2-ol
Molecular Weight (g/mol): 114.14
Molecular Formula: C6H10O2
InChi Key: GZCWLCBFPRFLKL-UHFFFAOYNA-N
SMILES: CC(O)COCC#C
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results