Learn More
Applied Biosystems™ Phosphalink™, for 3' and 5' Labeling, 70 mg
Catalog No. 401717
Encompass
Change view
Click to view available options
Quantity:
70 mg
Catalog No. Quantity
401717 70 mg
1 options
Catalog No. 401717 Supplier Applied Biosystems™ Supplier No. 401717
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Phosphalink™ is used for 3’ or 5’ attachment of phosphorous group during oligonucleotide synthesis.We offer several phosphoramidites that impart the means to add a phosphate during oligonucleotide synthesis.
Phosphate linker:Phosphalink™ phosphoramidite can incorporate a phosphate group at either the 5' or the 3' end. A 5'-end phosphate permits ligation of an oligonucleotide to other DNA.
  • Specialty phosphoramidites allow online addition of linkers during synthesis, saving time in production of labeled probes and primers.
  • Designed and optimized for use with ABI synthesizers, guaranteeing reliable nucleic acid synthesis.
  • We offer a range of specialty linkers and phosphoramites to give you the flexibility demanded in oligonucleotide synthesis.
Specifications
Packaging Type Shipped on Dry Ice
Quantity 70 mg
For Research Use Only. Not for use in diagnostic procedures.