missing translation for 'onlineSavingsMsg'
Learn More

Phloxine B, Spectrum™ Chemical

Catalog No. 18612304
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612304 10 g Amber Glass Bottle
18612306 25 g Amber Glass Bottle
18612305 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18612304 Supplier Spectrum Chemical Mfg Cor Supplier No. PH17010GM

Chemical Identifiers

CAS 18472-87-2

Specifications

CAS Max % 1
Grade Reagent
Packaging Amber Glass Bottle
Color Brown to Red
Quantity 10 g
Chemical Name or Material Phloxine B

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.