accessibility menu, dialog, popup

UserName

NaNaOBrOBrOBrOOClClClClBr

CAS RN 18472-87-2

IUPAC Name: disodium 2',4',5',7'-tetrabromo-4,5,6,7-tetrachloro-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthene]-3',6'-bis(olate)
Synonyms: phloxine acid red 92
Molecular Weight (g/mol): 829.63
Molecular Formula: C20H2Br4Cl4Na2O5
InChi Key: OOYIOIOOWUGAHD-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]C1=C(Br)C=C2C(OC3=C(Br)C([O-])=C(Br)C=C3C22OC(=O)C3=C(Cl)C(Cl)=C(Cl)C(Cl)=C23)=C1Br
Quantity 
  • (1)
  • (1)
  • (5)
  • (1)
Color 
  • (2)
  • (3)
  • (1)
  • (1)
Grade 
  • (3)
  • (3)
Percent Purity 
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (5)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)