Learn More
Thermo Scientific Chemicals Phloxine B, 85%, pure, high purity, Biological stain
CAS: 18472-87-2 | C20H2Br4Cl4Na2O5
$112.39 - $318.32
Chemical Identifiers
| CAS | 18472-87-2 |
|---|---|
| Molecular Formula | C20H2Br4Cl4Na2O5 |
| MDL Number | MFCD00005061 |
| Synonym | Acid Red 92, C.I. 45410 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC189470050
|
Thermo Scientific Chemicals
189470050 |
5 g | Glass bottle |
Each for $112.39
|
|
||||
|
AC189470250
|
Thermo Scientific Chemicals
189470250 |
25 g | Glass bottle |
Each for $318.32
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 18472-87-2 | |
| C20H2Br4Cl4Na2O5 | |
| MFCD00005061 | |
| Acid Red 92, C.I. 45410 |
Specifications
| 875 to 1300 (at lambda max.) | |
| 1.38 to 2.24 | |
| 85% | |
| MFCD00005061 | |
| 15, 7440 | |
| Authentic | |
| 10% max. (105°C) | |
| 85% | |
| Glass bottle | |
| 5 g | |
| Phloxine B |
| 18472-87-2 | |
| 80% min. (on anhydrous substance) | |
| C20H2Br4Cl4Na2O5 | |
| 19, IV, 2921 | |
| Acid Red 92, C.I. 45410 | |
| 546 to 550nm (in 50:50 METHS:water) | |
| 829.64 | |
| Pure | |
| Brown to Brown/Red | |
| Powder |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RUO – Research Use Only