missing translation for 'onlineSavingsMsg'
Learn More

Phloroglucinol Dihydrate 98.0+%, TCI America™

Catalog No. p-7135106
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P137625G 25 g
P1376100G 100 g
P1376500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P137625G Supplier TCI America Supplier No. P137625G
Only null left

Chemical Identifiers

CAS 6099-90-7
Molecular Formula C6H10O5
Molecular Weight (g/mol) 162.141
MDL Number MFCD00149090
InChI Key MPYXTIHPALVENR-UHFFFAOYSA-N
Synonym phloroglucinol dihydrate, benzene-1,3,5-triol dihydrate, 1,3,5-trihydroxybenzene dihydrate, 1,3,5-benzenetriol, dihydrate, phloroglucinol, dihydrate, sym-trihydroxybenze, unii-0x7b22brg1, ccris 6521, 1,3,5-benzoltrioldihydrat, phloroglucinoldihydrate
PubChem CID 80196
IUPAC Name benzene-1,3,5-triol;dihydrate
SMILES C1=C(C=C(C=C1O)O)O.O.O

Specifications

Melting Point 214°C
Quantity 25 g
Formula Weight 126.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Phloroglucinol Dihydrate
EINECSNumber (3)-0554
RTECSNumber DC4662500
TSCA No