Learn More
Phloroglucinol, 99+%, anhydrous
CAS: 108-73-6 | C6H6O3 | 126.1 g/mol
Supplier: Thermo Scientific Chemicals 241761000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Phloroglucinol | |
| 215.0°C to 218.0°C | |
| 100°C | |
| 99% min. (HPLC) | |
| C6H6O3 | |
| 100 g | |
| 15,7439 | |
| Solubility in water: 10g/L (20°C). Other solubilities: soluble in acetone ethanol, ether and pyridine | |
| C1=C(C=C(C=C1O)O)O | |
| 126.1 | |
| CHEBI:16204 | |
| 99+% |
| 108-73-6 | |
| White to Beige | |
| Authentic | |
| Glass bottle | |
| (OH)3C6H3 | |
| 01,47; 06,147 | |
| phloroglucinol, 1,3,5-benzenetriol, 1,3,5-trihydroxybenzene, phloroglucin, phloroglucine, spasfon-lyoc, s-trihydroxybenzene, benzene-s-triol, 5-hydroxyresorcinol, benzene, trihydroxy | |
| QCDYQQDYXPDABM-UHFFFAOYSA-N | |
| benzene-1,3,5-triol | |
| 359 | |
| 126.1 | |
| Crystalline Powder |
Chemical Identifiers
| 108-73-6 | |
| 126.1 | |
| phloroglucinol, 1,3,5-benzenetriol, 1,3,5-trihydroxybenzene, phloroglucin, phloroglucine, spasfon-lyoc, s-trihydroxybenzene, benzene-s-triol, 5-hydroxyresorcinol, benzene, trihydroxy | |
| CHEBI:16204 | |
| C1=C(C=C(C=C1O)O)O |
| C6H6O3 | |
| QCDYQQDYXPDABM-UHFFFAOYSA-N | |
| 359 | |
| benzene-1,3,5-triol |
Safety and Handling
GHS H Statement
Harmful to aquatic life with long lasting effects.
GHS P Statement
Avoid release to the environment.
Dispose of contents/container to an approved waste disposal plant.
Warning
EINECSNumber : 203-611-2
RUO – Research Use Only