missing translation for 'onlineSavingsMsg'
Learn More

PHKB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567288
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567288 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567288 Supplier Thermo Fisher Scientific Supplier No. PA567288
Only null left

PHKB Polyclonal Antibody

PHKB Polyclonal Antibody for ICC/IF

Specifications

Antigen PHKB
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PHKB
Gene Accession No. Q93100
Gene Alias AI463271; Phkb; Phosphorylase B; Phosphorylase b kinase regulatory subunit beta; phosphorylase kinase; phosphorylase kinase beta; phosphorylase kinase beta subunit; phosphorylase kinase beta-subunit; phosphorylase kinase regulatory subunit beta; phosphorylase kinase subunit beta; phosphorylase kinase, beta
Gene Symbols PHKB
Host Species Rabbit
Immunogen Recombinant Human PHKB. Recombinant protein control fragment (Product #RP-107926).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5257
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less