missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phire Reaction Buffers

Catalog No. F524L
Change view
Click to view available options
Chemical Name or Material:
Phire Green Reaction Buffer
Phire Reaction Buffer
Phire Reaction Buffer, detergent-free
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Chemical Name or Material
F524L Phire Reaction Buffer
F527L Phire Green Reaction Buffer
F525L Phire Reaction Buffer, detergent-free
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. F524L Supplier Thermo Scientific™ Supplier No. F524L

The reaction buffer for Thermo Scientific Phire Hot Start II DNA Polymerase is available in three formats: non-colored version with and without detergents or Green version containing a density reagent and two tracking dyes for direct loading of PCR products on gel.

Thermo Scientific Phire Reaction Buffer is designed for Phire Hot Start II DNA Polymerase. This 5X buffer solution is available in three formats, a non-colored version with or without detergents and a green version that contains a density reagent and two tracking dyes for direct loading of PCR products on gel. The colored buffer does not interfere with enzyme performance and is compatible with downstream applications such as DNA sequencing, ligation, and restriction digestion.

Phire Reaction Green Buffer includes a blue die that migrates with 3–5 kb DNA fragments in 1% agarose gel and a yellow dye that migrates faster than 10 bp DNA fragments in 1% agarose gel. The dyes have excitation peaks at 424 nm and 615 nm.

The concentration of Mg2+ in Phire Reaction Buffer is optimized to work well for most amplicons. The buffer contains 1.5 mM MgCl2 at final reaction concentration. Separate tubes of MgCl2 and DMSO are provided for further optimization.

Notes

  • For applications that require PCR product analysis by absorbance or fluorescence excitation, the colorless Phire Reaction Buffer is recommended.

Specifications

Chemical Name or Material Phire Reaction Buffer
Description Colorless
Concentration 5X
For Use With (Application) Standard PCR
Quantity 4 x 1.5 mL
Includes • 5X Phire Reaction Buffer, 4 x 1.5 mL
• 50 mM MgCl2, 2 x 1.5 mL
• DMSO, 0.5 mL

Store at -20°C.
Can I use Phusion Green and Phire Green PCR products for subsequent applications such as sequencing, ligation, and restriction digestion?

Yes. The dyes do not interfere with downstream applications such as DNA sequencing, ligation, and restriction digestion.

What dyes are present in Phusion Green and Phire Green buffers?

Phusion Green and Phire Green buffers contain two dyes for monitoring electrophoresis progress. The blue dye migrates with 3-5 kb DNA fragments and the yellow dye migrates faster than 10 bp DNA fragments in 1% agarose gel. The dyes have excitation peaks at 424 nm and 615 nm, respectively.

For Research Use Only. Not for use in diagnostic procedures.