Learn More
Bulbtronics™ Philips™ Linear Double-Ended Germicidal Lamps
Wavelength: 254nm
Supplier: Bulbtronics™ BBT233
Chemical Identifiers
| Germicidal UV-C | |
| 254 nm | |
| Linear Double-ended Lamp |
Specifications
| Medium Bi-pin (G13) | |
| 8000 hr. | |
| 254 nm | |
| 2.5 cm | |
| 36 in. | |
| T8 |
| Germicidal UV-C | |
| 30 W | |
| 1 in. | |
| 36 x 1 in. (91.4 x 2.5 cm) | |
| 91.4 cm | |
| Linear Double-ended Lamp |