missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ phi29 DNA Polymerase (10 U/μL)

Catalog No. FEREP0094
Change view
Click to view available options
Quantity:
1,000 units
250 units
5,000 units
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEREP0094 5,000 units
FEREP0091 250 units
FEREP0092 1,000 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. FEREP0094 Supplier Thermo Scientific™ Supplier No. EP0094

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific phi29 DNA Polymerase is a highly processive polymerase featuring strong strand displacement activity, which allows for highly efficient isothermal DNA amplification.

Thermo Scientific phi29 DNA Polymerase is a highly processive polymerase featuring strong strand displacement activity, which allows for highly efficient isothermal DNA amplification. Produced from E. coli cells with a cloned gene 2 of Bacillus subtilis phage phi29, phi29 DNA Polymerase possesses 3'→5' proofreading exonuclease activity that acts preferentially on single-stranded DNA or RNA.

Thermo Scientific phi29 DNA Polymerase is a highly processive polymerase featuring strong strand displacement activity, which allows for highly efficient isothermal DNA amplification. Produced from E. coli cells with a cloned gene 2 of Bacillus subtilis phage phi29, phi29 DNA Polymerase possesses 3'→5' proofreading exonuclease activity that acts preferentially on single-stranded DNA or RNA.

Features of phi29 DNA Polymerase
• Excellent processivity and strand displacement activity—more than 70 kb long DNA stretches can be synthesized
• Highly accurate DNA synthesis
• High yields of amplified DNA even from small amounts of template

Applications
Phi29 DNA Polymerase may be used in a wide variety of applications including:
• Rolling circle amplification (RCA)—generation of periodic DNA nanotemplates
• Multiple displacement amplification (MDA)
• Unbiased amplification of whole genome (WGA) (see figure below)
• Amplification of DNA for SNP and STR detection
• Cell-free amplification of DNA from single cells, pathogenic organisms, or metagenomes
• DNA template preparation for sequencing
• Protein-primed DNA amplification
• Recombination based-cloning
• Cell-free cloning of lethal DNA
• RNA-primed DNA amplification

Notes
1. The addition of pyrophosphatase to the reaction mixture with phi29 DNA Polymerase may enhance DNA synthesis.
2. Because of the enzymes’s 3’→5’ proofreading exonuclease, 3'-modified primers are highly recommended.

Specifications

Product Type Stand-alone enzyme
For Use With (Application) MDA-WGA, RCA
Polymerase phi29 DNA Polymerase
Quantity 5,000 units
Content And Storage • phi29 DNA Polymerase (5,000 units at 10 U/μL)
• 10X Reaction Buffer for phi29 DNA Polymerase
• Store at –15 to –25°C.
Optimal Reaction Temperature 30°C
Shipping Condition Dry Ice
Concentration 10 U/μL
Reaction Time 4 hours
What is the error rate of phi29 DNA Polymerase?

The error rate of phi29 DNA Polymerase is 1 x 10-5.
The error rate of phi29 DNA Polymerase was measured according to the method described in literature:
Mielinis, P., Sukackaitė, R., Serapinaitė, A., Samoilovas, F., Alzbutas, G., Matjošaitis, K., & Lubys, A. (2021). MUA-based molecular indexing for rare mutation detection by Next-Generation sequencing. Journal of Molecular Biology, 433(19), 167209. https://doi.org/10.1016/j.jmb.2021.167209

Can I use phi29 DNA Polymerase to incorporate 5-methyl-dCTP?

phi29 DNA Polymerase should be able to incorporate 5-methyl-dCTP nucleotides and other modified nucleotides.
The incorporation of modified nucleotides by phi29 DNA Polymerase can be reviewed in the literature:
Jakubovska, J., Tauraitė, D., Birštonas, L., & Meškys, R. (2018). N 4-acyl-2′-deoxycytidine-5′-triphosphates for the enzymatic synthesis of modified DNA. Nucleic Acids Research, 46(12), 5911–5923. https://doi.org/10.1093/nar/gky435

What is the minimal recommended time for amplification with phi29 DNA Polymerase?

The optimal reaction time for DNA amplification with phi29 DNA Polymerase is 4 hours. For samples with ≥1 pg of DNA input, DNA amplification time can be shortened to 2 hours if maximizing product yield is not essential.

Can phi29 DNA Polymerase be used with liquid media culture or colonies?

Yes. phi29 DNA Polymerase can work with different types of sample input such as purified DNA, liquid media culture, agar plate colonies, etc.

Can phi29 DNA Polymerase be heat-inactivated?

Yes, we recommend heat-inactivating phi29 DNA Polymerase at 65 degrees C for 10 min.

Is phi29 DNA Polymerase supplied with dNTPs?

No. dNTPs can be purchased separately (Cat. No. R0181).

Is phi29/EquiPhi29 DNA Polymerase suitable for amplification of short DNA fragments?

The amplification efficiency of MDA (multiple displacement amplification) reaction rapidly diminishes as the molecular weight of the starting material decreases, thus making the polymerase unsuitable for amplification of low-molecular weight DNA. However, it is suitable for amplification from extremely low amounts of starting DNA material, making it ideal for single cell analysis.

What type of DNA template can be amplified by phi29 DNA Polymerase?

Both linear and circular DNA templates can be amplified by phi29 DNA Polymerase.


For Research Use Only. Not for use in diagnostic procedures.