missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PHF20 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA527878
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA527878 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA527878 Supplier Invitrogen™ Supplier No. PA527878

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, Molt-4, Raji. Predicted reactivity: Mouse (95%), Rat (93%), Rabbit (94%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Methyllysine-binding protein, component of the MOF histone acetyltransferase protein complex. Not required for maintaining the global histone H4 'Lys-16' acetylation (H4K16ac) levels or locus specific histone acetylation, but instead works downstream in transcriptional regulation of MOF target genes. As part of the NSL complex it may be involved in acetylation of nucleosomal histone H4 on several lysine residues. Contributes to methyllysine-dependent p53/TP53 stabilization and up-regulation after DNA damage.

Specifications

Antigen PHF20
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene PHF20
Gene Accession No. Q9BVI0
Gene Alias 6820402O20Rik; C20orf104; GLEA2; Glioma-expressed antigen 2; HCA58; Hepatocellular carcinoma-associated antigen 58; hepatocellular carcinoma-associated antigen 58 homolog; Novel zinc finger protein; NZF; PHD finger 20-like protein; PHD finger protein 20; PHD finger protein 20-like protein; Phf20; TDRD20A; transcription factor TZP; tudor domain containing 20A; TZP
Gene Symbols PHF20
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human PHF20. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51230
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less