missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PHF10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112648
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112648 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112648 Supplier Invitrogen™ Supplier No. PA5112648
Only null left

Rabbit Polyclonal Antibody

PHF10 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

This gene contains a predicted ORF that encodes a protein with two zinc finger domains. The function of the encoded protein is not known. Sequence analysis suggests that multiple alternatively spliced transcript variants are derived from this gene but the full-length nature of only two of them is known. These two splice variants encode different isoforms. A pseudogene for this gene is located on Xq28.

Specifications

Antigen PHF10
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PHF10
Gene Accession No. Q8WUB8
Gene Alias 1810055P05Rik; AV024533; BAF45A; BRG1-associated factor 45a; BRG1-associated factor, 45-KD, A; PHD finger protein 10; PHD finger protein 10, isoform a; PHD zinc finger protein XAP135; Phf10; XAP135
Gene Symbols PHF10
Host Species Rabbit
Immunogen Recombinant Human PHD finger protein 10 protein (151-360AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55274
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less