missing translation for 'onlineSavingsMsg'
Learn More

Phenytoin sodium Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111017854
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017854 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017854 Supplier Merck Emd Millipore Supplier No. PHR14921G

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 630-93-3
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C15H11N2O2 Na
MDL Number MFCD00069674
Synonym 5,5-Diphenylhydantoin sodium salt; 5,5-Diphenyl-2,4-imidazolidinedione; Phenytoin
Type Certified Reference Material
Quantity 1 g
Formula Weight 274.25
Grade Certified Reference Material