missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenyltrimethylammonium hydroxide, 0.1 M in methanol
CAS: 1899-02-1 | C9H15NO | 153.23 g/mol
Supplier: Thermo Scientific Chemicals 422021000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalystSpecifications
| Phenyltrimethylammonium hydroxide | |
| 0.1M in Methanol | |
| C9H15NO | |
| MFCD00041899 | |
| 0.8000g/mL | |
| HADKRTWCOYPCPH-UHFFFAOYSA-M | |
| N,N,N-trimethylanilinium hydroxide | |
| 15913 | |
| 153.22 | |
| 0.8 | |
| 10°C |
| Colorless | |
| 1899-02-1,67-56-1 | |
| C6H5N(CH3)3OH | |
| 0.09 to 0.11M | |
| methelute, phenyltrimethylammonium hydroxide, trimethylanilinium hydroxide, phenyl trimethyl ammonium hydroxide, trimethylphenylammonium hydroxide, n,n,n-trimethylbenzenaminium hydroxide, n,n,n-trimethylanilinium hydroxide, benzenaminium, n,n,n-trimethyl-, hydroxide, ptah, ammonium, phenyltrimethyl-, hydroxide | |
| [OH-].C[N+](C)(C)C1=CC=CC=C1 | |
| 153.23 | |
| CHEBI:85062 | |
| Glass bottle | |
| 100 g | |
| Liquid |
RUO – Research Use Only