missing translation for 'onlineSavingsMsg'
Learn More

Phenyltrimethoxysilane, 85%, Thermo Scientific™

$101.83 - $346.55

Chemical Identifiers

CAS 2996-92-1
Molecular Formula C9H14O3Si
Molecular Weight (g/mol) 198.29
MDL Number MFCD00025689
InChI Key ZNOCGWVLWPVKAO-UHFFFAOYSA-N
Synonym phenyltrimethoxysilane, trimethoxy phenyl silane, silane, trimethoxyphenyl, silane, phenyltrimethoxy, unii-21tqe746s9, trimethoxysilyl benzene, benzene, trimethoxysilyl, phenyl trimethoxysilane, phenyl trimethoxy silane, phenyltri methoxy silane
PubChem CID 18137
IUPAC Name trimethoxy(phenyl)silane
SMILES CO[Si](OC)(OC)C1=CC=CC=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC370640500
View Documents Promotion Details
N/A 50mL Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC370642500
View Documents Promotion Details
N/A 250mL Glass bottle
Each for $101.83
Only null left
 
AC370640010
View Documents Promotion Details
N/A 1L Glass bottle
Each for $346.55
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

2996-92-1
198.29
ZNOCGWVLWPVKAO-UHFFFAOYSA-N
18137
CO[Si](OC)(OC)C1=CC=CC=C1
C9H14O3Si
MFCD00025689
phenyltrimethoxysilane, trimethoxy phenyl silane, silane, trimethoxyphenyl, silane, phenyltrimethoxy, unii-21tqe746s9, trimethoxysilyl benzene, benzene, trimethoxysilyl, phenyl trimethoxysilane, phenyl trimethoxy silane, phenyltri methoxy silane
trimethoxy(phenyl)silane

Specifications

2996-92-1
233°C
Authentic
C9H14O3Si
C6H5Si(OCH3)3
16, IV, 1556
phenyltrimethoxysilane, trimethoxy phenyl silane, silane, trimethoxyphenyl, silane, phenyltrimethoxy, unii-21tqe746s9, trimethoxysilyl benzene, benzene, trimethoxysilyl, phenyl trimethoxysilane, phenyl trimethoxy silane, phenyltri methoxy silane
ZNOCGWVLWPVKAO-UHFFFAOYSA-N
trimethoxy(phenyl)silane
18137
85%
1.0620g/mL
50°C
80% min. (GC)
1.4730 to 1.475
MFCD00025689
1.062
Solubility in water: soluble
CO[Si](OC)(OC)C1=CC=CC=C1
198.29
198.29
Phenyltrimethoxysilane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure if swallowed.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: rinse mouth.
Do NOT induce vomi

GHS Signal Word: Warning

EINECSNumber : 221-066-9

SDS
Documents

Documents

RUO – Research Use Only