missing translation for 'onlineSavingsMsg'
Learn More

Phenyltrichlorosilane 98.0+%, TCI America™

Catalog No. p-7134512
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P024025G 25 g
P0240500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P024025G Supplier TCI America Supplier No. P024025G

Chemical Identifiers

CAS 98-13-5
Molecular Formula C6H5Cl3Si
Molecular Weight (g/mol) 211.541
MDL Number MFCD00000479
InChI Key ORVMIVQULIKXCP-UHFFFAOYSA-N
Synonym trichloro phenyl silane, phenyltrichlorosilane, silane, trichlorophenyl, phenyl trichlorosilane, phenylsilicon trichloride, silane, phenyltrichloro, silicon phenyl trichloride, benzene, trichlorosilyl, unii-m7199jl06m, phenyl-trichloro-silane
PubChem CID 7372
IUPAC Name trichloro(phenyl)silane
SMILES C1=CC=C(C=C1)[Si](Cl)(Cl)Cl

Specifications

Color Colorless
Boiling Point 201°C
Quantity 25 g
UN Number 1804
Formula Weight 211.54
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Phenyltrichlorosilane
EINECSNumber (3)-2634
RTECSNumber VV6650000
TSCA Yes