accessibility menu, dialog, popup

UserName

CAS RN 98-13-5

IUPAC Name: trichloro(phenyl)silane
Synonyms: phenyltrichlorosilane
Molecular Weight (g/mol): 211.54
Molecular Formula: C6H5Cl3Si
InChi Key: ORVMIVQULIKXCP-UHFFFAOYSA-N
SMILES: Cl[Si](Cl)(Cl)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (4)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (5)