missing translation for 'onlineSavingsMsg'
Learn More

Phenyltrichlorosilane, 97%

Catalog No. p-7023301
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1671322 100 g
AAA1671336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1671322 Supplier Thermo Scientific Chemicals Supplier No. A1671322

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 98-13-5 | C6H5Cl3Si | 211.541 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 98-13-5
Molecular Formula C6H5Cl3Si
Molecular Weight (g/mol) 211.541
MDL Number MFCD00000479
InChI Key ORVMIVQULIKXCP-UHFFFAOYSA-N
Synonym trichloro phenyl silane, phenyltrichlorosilane, silane, trichlorophenyl, phenyl trichlorosilane, phenylsilicon trichloride, silane, phenyltrichloro, silicon phenyl trichloride, benzene, trichlorosilyl, unii-m7199jl06m, phenyl-trichloro-silane
PubChem CID 7372
IUPAC Name trichloro(phenyl)silane
SMILES C1=CC=C(C=C1)[Si](Cl)(Cl)Cl

Specifications

Boiling Point 201°C
Melting Point -127°C
Quantity 100 g
Flash Point 91°C (195°F)
Odor Odorless
Refractive Index 1.523
Sensitivity Moisture sensitive
Density 1.324
Linear Formula (C6H5)SiCl3
UN Number UN1804
Beilstein 508730
Formula Weight 211.55
Percent Purity 97%
Chemical Name or Material Phenyltrichlorosilane
Show More Show Less
Hazard Category H227-H311-H314
Hazard Statement GHS H Statement
H330-H314-H318-H227-H312
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Harmful in contact with skin.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: PHENYLTRICHLOROSILANE
EINECSNumber 202-640-8
RTECSNumber VV6650000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only