missing translation for 'onlineSavingsMsg'
Learn More

Phenyltrichlorosilane, 95%

CAS: 98-13-5 | C6H5Cl3Si | 211.55 g/mol

$145.36 - $145.36

Chemical Identifiers

CAS 98-13-5
Molecular Formula C6H5Cl3Si
Molecular Weight (g/mol) 211.55
MDL Number MFCD00000479
InChI Key ORVMIVQULIKXCP-UHFFFAOYSA-N
Synonym trichloro phenyl silane, phenyltrichlorosilane, silane, trichlorophenyl, phenyl trichlorosilane, phenylsilicon trichloride, silane, phenyltrichloro, silicon phenyl trichloride, benzene, trichlorosilyl, unii-m7199jl06m, phenyl-trichloro-silane
PubChem CID 7372
IUPAC Name trichloro(phenyl)silane
SMILES C1=CC=C(C=C1)[Si](Cl)(Cl)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC131002500
SDS View Documents
Thermo Scientific Chemicals
131002500
250 mL Glass bottle
Each for $145.36
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

98-13-5
211.55
ORVMIVQULIKXCP-UHFFFAOYSA-N
7372
C1=CC=C(C=C1)[Si](Cl)(Cl)Cl
C6H5Cl3Si
MFCD00000479
trichloro phenyl silane, phenyltrichlorosilane, silane, trichlorophenyl, phenyl trichlorosilane, phenylsilicon trichloride, silane, phenyltrichloro, silicon phenyl trichloride, benzene, trichlorosilyl, unii-m7199jl06m, phenyl-trichloro-silane
trichloro(phenyl)silane

Specifications

98-13-5
1.3210g/mL
91°C
94% min. (GC)
C6H5Cl3Si
C6H5SiCl3
250 mL
1.321
Solubility in water: reacts. Other solubilities: soluble in benzene,ehter and perchloroethylene
C1=CC=C(C=C1)[Si](Cl)(Cl)Cl
211.55
211.55
Liquid
Colorless to Yellow
201°C
Authentic
Glass bottle
1.5220 to 1.524
MFCD00000479
16, 911
trichloro phenyl silane, phenyltrichlorosilane, silane, trichlorophenyl, phenyl trichlorosilane, phenylsilicon trichloride, silane, phenyltrichloro, silicon phenyl trichloride, benzene, trichlorosilyl, unii-m7199jl06m, phenyl-trichloro-silane
ORVMIVQULIKXCP-UHFFFAOYSA-N
trichloro(phenyl)silane
7372
95%
Phenyltrichlorosilane, 95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Reacts violently with water.
Corrosive to the respiratory tract.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately

GHS Signal Word: Danger

EINECSNumber : 202-640-8

RTECSNumber : VV6650000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only