missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenylthiohydantoin-proline 98.0+%, TCI America™
$102.52 - $667.33
Chemical Identifiers
| CAS | 4333-21-5 |
|---|---|
| Molecular Formula | C12H12N2OS |
| Molecular Weight (g/mol) | 232.301 |
| MDL Number | MFCD00056895 |
| InChI Key | YNJJBMAKBUZCKN-UHFFFAOYSA-N |
| Synonym | PTH-proline |
| PubChem CID | 100798 |
| IUPAC Name | 2-phenyl-3-sulfanylidene-5,6,7,7a-tetrahydropyrrolo[1,2-c]imidazol-1-one |
| SMILES | C1CC2C(=O)N(C(=S)N2C1)C3=CC=CC=C3 |
Chemical Identifiers
| 4333-21-5 | |
| 232.301 | |
| YNJJBMAKBUZCKN-UHFFFAOYSA-N | |
| 100798 | |
| C1CC2C(=O)N(C(=S)N2C1)C3=CC=CC=C3 |
| C12H12N2OS | |
| MFCD00056895 | |
| PTH-proline | |
| 2-phenyl-3-sulfanylidene-5,6,7,7a-tetrahydropyrrolo[1,2-c]imidazol-1-one |
Specifications
| 4333-21-5 | |
| White | |
| MFCD00056895 | |
| PTH-proline | |
| C1CC2C(=O)N(C(=S)N2C1)C3=CC=CC=C3 | |
| 232.301 | |
| 232.30 | |
| Crystalline Powder |
| 179°C | |
| C12H12N2OS | |
| 100 mg | |
| YNJJBMAKBUZCKN-UHFFFAOYSA-N | |
| 2-phenyl-3-sulfanylidene-5,6,7,7a-tetrahydropyrrolo[1,2-c]imidazol-1-one | |
| 100798 | |
| ≥98.0% (HPLC,N) | |
| Phenylthiohydantoin-proline |
Safety and Handling
TSCA : No