missing translation for 'onlineSavingsMsg'
Learn More

Phenylthiohydantoin-methionine 98.0+%, TCI America™

Catalog No. P0366100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0366100MG 100 mg
P03661G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0366100MG Supplier TCI America Supplier No. P0366100MG
Only null left

Chemical Identifiers

CAS 4370-90-5
Molecular Formula C12H14N2OS2
Molecular Weight (g/mol) 266.377
MDL Number MFCD06858379
InChI Key KALSXWPXUWRLOH-UHFFFAOYSA-N
Synonym 5-[2-(Methylthio)ethyl]-3-phenyl-2-thiohydantoin, PTH-methionine
PubChem CID 2872023
IUPAC Name 5-(2-methylsulfanylethyl)-3-phenyl-2-sulfanylideneimidazolidin-4-one
SMILES CSCCC1C(=O)N(C(=S)N1)C2=CC=CC=C2

Specifications

Melting Point 133°C
Color White
Quantity 100 mg
Formula Weight 266.38
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Phenylthiohydantoin-methionine
RTECSNumber MU2700000
TSCA No