accessibility menu, dialog, popup

UserName

CH3SNHSNO

CAS RN 4370-90-5

IUPAC Name: 5-[2-(methylsulfanyl)ethyl]-3-phenyl-2-sulfanylideneimidazolidin-4-one
Synonyms: 5-[2-(methylsulfanyl)ethyl]-3-phenyl-2-sulfanylideneimidazolidin-4-one
Molecular Weight (g/mol): 266.38
Molecular Formula: C12H14N2OS2
InChi Key: KALSXWPXUWRLOH-UHFFFAOYNA-N
SMILES: CSCCC1NC(=S)N(C1=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results