missing translation for 'onlineSavingsMsg'
Learn More

Phenylthiohydantoin-aspartic Acid 95.0+%, TCI America™

Catalog No. p-7134572
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0364100MG 100 mg
P03641G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0364100MG Supplier TCI America Supplier No. P0364100MG

Chemical Identifiers

CAS 5624-13-5
Molecular Formula C11H10N2O3S
Molecular Weight (g/mol) 250.272
MDL Number MFCD01076622
InChI Key NFUCNAREVSTFNC-UHFFFAOYSA-N
Synonym 3-Phenyl-2-thiohydantoin-5-acetic Acid, PTH-aspartic Acid
PubChem CID 2871997
IUPAC Name 2-(5-oxo-1-phenyl-2-sulfanylideneimidazolidin-4-yl)acetic acid
SMILES C1=CC=C(C=C1)N2C(=O)C(NC2=S)CC(=O)O

Specifications

Color White-Yellow
Quantity 100 mg
Formula Weight 250.27
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Phenylthiohydantoin-aspartic Acid
RTECSNumber NI8720800
TSCA No