accessibility menu, dialog, popup

UserName

CAS RN 5624-13-5

IUPAC Name: 2-(5-oxo-1-phenyl-2-sulfanylideneimidazolidin-4-yl)acetic acid
Synonyms: aspartic phenylthiohydantoin
Molecular Weight (g/mol): 250.27
Molecular Formula: C11H10N2O3S
InChi Key: NFUCNAREVSTFNC-UHFFFAOYNA-N
SMILES: OC(=O)CC1NC(=S)N(C1=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results