Learn More
(Phenylsulfonyl)acetic acid, 97%
CAS: 3959-23-7 | C8H8O4S | 200.208 g/mol
Supplier: Thermo Scientific Chemicals L0829906
Description
(Phenylsulfonyl)acetic acid is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(Phenylsulfonyl)acetic acid is used as pharmaceutical intermediates.
Notes
Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place
Specifications
| (Phenylsulfonyl)acetic acid | |
| 109°C to 114°C | |
| MFCD00025042 | |
| 2504376 | |
| YTEFAALYDTWTLB-UHFFFAOYSA-N | |
| 2-(benzenesulfonyl)acetic acid | |
| 59543 | |
| 97% |
| 3959-23-7 | |
| C8H8O4S | |
| 5 g | |
| phenylsulfonyl acetic acid, 2-phenylsulfonyl acetic acid, 2-benzenesulfonyl acetic acid, phenylsulfonylacetic acid, acetic acid, phenylsulfonyl, benzenesulfonyl-acetic acid, benzenesulfonyl acetic acid, 2-phenylsulphonyl acetic acid, benzenesulfonylacetic acid, phenyl sulfonylacetic acid | |
| C1=CC=C(C=C1)S(=O)(=O)CC(=O)O | |
| 200.208 | |
| 200.22 |
Chemical Identifiers
| 3959-23-7 | |
| 200.208 | |
| YTEFAALYDTWTLB-UHFFFAOYSA-N | |
| 59543 | |
| C1=CC=C(C=C1)S(=O)(=O)CC(=O)O |
| C8H8O4S | |
| MFCD00025042 | |
| phenylsulfonyl acetic acid, 2-phenylsulfonyl acetic acid, 2-benzenesulfonyl acetic acid, phenylsulfonylacetic acid, acetic acid, phenylsulfonyl, benzenesulfonyl-acetic acid, benzenesulfonyl acetic acid, 2-phenylsulphonyl acetic acid, benzenesulfonylacetic acid, phenyl sulfonylacetic acid | |
| 2-(benzenesulfonyl)acetic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only