missing translation for 'onlineSavingsMsg'
Learn More

(Phenylsulfonyl)acetic acid, 97%

Catalog No. AAL0829914
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0829914 25 g
AAL0829903 1 g
AAL0829906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0829914 Supplier Thermo Scientific Chemicals Supplier No. L0829914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3959-23-7 | C8H8O4S | 200.208 g/mol

(Phenylsulfonyl)acetic acid is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Phenylsulfonyl)acetic acid is used as pharmaceutical intermediates.

Notes
Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place

Chemical Identifiers

CAS 3959-23-7
Molecular Formula C8H8O4S
Molecular Weight (g/mol) 200.208
MDL Number MFCD00025042
InChI Key YTEFAALYDTWTLB-UHFFFAOYSA-N
Synonym phenylsulfonyl acetic acid, 2-phenylsulfonyl acetic acid, 2-benzenesulfonyl acetic acid, phenylsulfonylacetic acid, acetic acid, phenylsulfonyl, benzenesulfonyl-acetic acid, benzenesulfonyl acetic acid, 2-phenylsulphonyl acetic acid, benzenesulfonylacetic acid, phenyl sulfonylacetic acid
PubChem CID 59543
IUPAC Name 2-(benzenesulfonyl)acetic acid
SMILES C1=CC=C(C=C1)S(=O)(=O)CC(=O)O

Specifications

Melting Point 109°C to 114°C
Quantity 25 g
Beilstein 2504376
Formula Weight 200.22
Percent Purity 97%
Chemical Name or Material (Phenylsulfonyl)acetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only