missing translation for 'onlineSavingsMsg'
Learn More

Phenylsilane 97.0+%, TCI America™

Supplier:  TCI America P12915ML

 View more versions of this product

Catalog No. P12915ML

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Phenylsilane
Yellow
C6H5Si
5 mL
phenylsilane, benzene, silyl, silylbenzene, silane, phenyl, fenylsilan, fenylsilan czech, phenylsilane, silyl, silane, phenyl-,
C1=CC=C(C=C1)[Si]
105.191
108.22
Liquid
694-53-1
120°C
MFCD00013478
1993
XJWOWXZSFTXJEX-UHFFFAOYSA-N
phenylsilicon
6327628
≥97.0% (GC)

Chemical Identifiers

694-53-1
105.191
XJWOWXZSFTXJEX-UHFFFAOYSA-N
6327628
C1=CC=C(C=C1)[Si]
C6H5Si
MFCD00013478
phenylsilane, benzene, silyl, silylbenzene, silane, phenyl, fenylsilan, fenylsilan czech, phenylsilane, silyl, silane, phenyl-,
phenylsilicon
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : VV4825000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions